Orders LinkContactUsLinkComponent
Automatic burettes, self-zeroing, Nalgene®
Automatic burettes, self-zeroing, Nalgene®
Automatic burettes, self-zeroing, Nalgene®
Automatic burettes, self-zeroing, Nalgene®
Crystal-clear acrylic, PMP tip and TFE stopcock and plug, LDPE bottle with PP screw closure, 1 m PVC tubing.
  • Unaffected by dilute mineral acids and bases except HF and NH₄OH
  • Not designed for alcohol and organic solvents
Durable, excellent for general use, schools, industrial labs and field use.
ASTM E287
 : ASTM E287
  • Catalog No:
  • 612-2583
  • 612-2584
  • 612-2585
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Certifikát ochrany životního prostředí:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Kapacita
    Dělení
    Tolerance
  • 10 ml
    0,1 ml
    0,01 ± ml
    1 * 1 KS
    1 * 1 KS - 5 730,00 Kč
    5 730,00 Kč
    1 * 1 KS
       5 730,00 Kč
      Low Manufacturing Impact
    Specifications
    Cat. No.
    612-2583
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 25 ml
    0,1 ml
    0,01 ± ml
    1 * 1 KS
    1 * 1 KS - 6 100,00 Kč
    6 100,00 Kč
    1 * 1 KS
       6 100,00 Kč
      Low Manufacturing Impact
    Specifications
    Cat. No.
    612-2584
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 ml
    0,1 ml
    0,01 ± ml
    1 * 1 KS
    1 * 1 KS - 6 900,00 Kč
    6 900,00 Kč
    1 * 1 KS
       6 900,00 Kč
      Low Manufacturing Impact
    Specifications
    Cat. No.
    612-2585
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
  •  
    Popis
  • NALG71-3650-1025
    Stopcock assembly for burette, 25 ml
    1 * 2 KS - 8 290,00 Kč