Quick Chill™ Cooling unit, Nalgene®
Catalog # 479-5160
Supplier: Thermo Fisher Scientific
Specifications
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Bal.:1
Specifications
About this item
PC, non-toxic insulating solution.
- No direct contact between tubes and insulating solution so labels and markings won't come off
- Stackable and have non-slip rubber feet
For rapid chilling of biological samples. Can be used repeatedly for up to 45 minutes. It will chill up to 2 ml of alcohol-based solutions to −20 °C in less than 3 minutes or up to 2 ml of water-based solutions to 0 °C in less than 4 minutes.
For 12 (3×4) microtubes of 1,5 or 0,5 ml.
W×D×H: 151×108×125 mm