Conical-bottom centrifuge tube adapters, Nalgene®
525-0143
- Ø:28,83 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:6
PC, autoclavable.
- These adapters must be used with Nalgene® conical-bottom centrifuge tubes (35 ml)
- Fits most commonly used low- and high-speed rotors
- Easily removed from rotors after use