Replacement stopcock for aspirator bottles, Azlon®
Catalog # 215-3164
Supplier: SciLabware
Specifications
- Popis:Replacement stopcock for octagonal aspirator bottles
- Bal.:1
Specifications
About this item
PP, heavy-duty.
- Very good chemical resistance
- Leakproof
Boss-fitting stopcock is suitable for carboys/aspirator bottles 215-3161 to 215-3163.