Replacement spigot for carboys and aspirator bottles
227-4009
: KARTELL
- Popis:Replacement stopcock for carboys/aspirator bottles
- Bal.:1
HDPE, white.
- Good chemical resistance
- Leakproof
- Suitable for carboys/aspirator bottles 216-5744 to 216-5747
With ¾" BSP screw thread.