Media bottle closures, Top Works™ system, Nalgene®
Katalogové číslo 700-0001
Dodavatel: Thermo Fisher Scientific
Specifikace
- Připojení:2x6,35 mm/1x3,2 mm
- Závit:GL 45
- Environmentally Preferable:Low Manufacturing Impact
- Bal.:1
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
Specifikace
O tomto produktu
PSF, closures with platinum cured silicone inserts.
- They fit the corresponding threads on bottles by Schott, Corning and Wheaton
- For thread size GL 45
- Autoclavable, leakproof
These closures make it easier to transfer liquids aseptically from most conventional glass media bottles.