Aspirator bottles with screw cap and spigot
215-0969
: MURRAY S
- Kapacita:10000 ml
- Sekvence:2
- Bal.:1
HDPE, translucent with red screw cap.
- Good chemical resistance
- Heat resistant up to +80 °C (+120 °C for short periods)
- Thick walled, very robust
With tightly fitting spigot. Ideal for storing and dispensing liquids. Also recommended for use as settling bottles. Equipped with a carrying handle.