ContactUsLinkComponent
Measuring beaker with handle, Azlon®
Measuring beaker with handle, Azlon®
Catalog # 213-0154
Supplier:  SciLabware
Supplier #: JPM0250P
undefined
Measuring beaker with handle, Azlon®
Catalog # 213-0154
Supplier:  SciLabware
Supplier #: JPM0250P
Log in for availability
Select an Option
70 mm
Select an Option
50 ml
Select an Option
250 ml
Select an Option
Select an Option
120 mm
Select an Option
03
Select an Option
1
About this item
PP, highly transparent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. Clear moulded graduations. Ergonomically designed handle with thumb grip for easy carrying.
Frequently Bought Together