ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # 213-0376
Supplier:  RSG
Supplier #: 077.500
undefined
Measuring beakers with handle
Catalog # 213-0376
Supplier:  RSG
Supplier #: 077.500
Log in for availability
Select an Option
80 mm
Select an Option
500 ml
Select an Option
Select an Option
110 mm
Select an Option
03
Select an Option
1
About this item
Stainless steel, rust-proof, with flared rim, spout and handle.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Frequently Bought Together