ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # 213-3730
Supplier:  VITLAB
Supplier #: 443081
undefined
Measuring beakers with handle
Catalog # 213-3730
Supplier:  VITLAB
Supplier #: 443081
Log in for availability
Select an Option
152 mm
Select an Option
20 ml
Select an Option
2000 ml
Select an Option
Select an Option
213 mm
Select an Option
06
Select an Option
1
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products acc. to EC Directive No. 10/2011.
Frequently Bought Together