ContactUsLinkComponent
Measuring beakers with handle, Nalgene®
  213-7172
 : DS1511-1000
Low Manufacturing Impact
undefined
Measuring beakers with handle, Nalgene®
  213-7172
 : DS1511-1000
  Low Manufacturing Impact
Select an Option
1000 ml
Select an Option
Select an Option
01
Select an Option
1
  • Catalog No:
  • 213-7172
  • 213-7174
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Certifikát ochrany životního prostředí:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
PFA, translucent.
  • Exceptional chemical resistance
  • Very good thermal resistance (−270 to +250 °C), autoclavable
  • Very sturdy, extra strong walls
These beakers are exceptionally chemically resistant and can be used repeatedly for ultrapure applications. For use with strong oxidising agents, acids, hydrocarbons and ketones. The material is non-adhesive, so the measuring beakers are easy to clean. With spout.
Frequently Bought Together