ContactUsLinkComponent
Beakers, low form
Beakers, low form
Catalog # 213-1911
Supplier:  KARTELL
Supplier #: 1720
undefined
Beakers, low form
Catalog # 213-1911
Supplier:  KARTELL
Supplier #: 1720
Log in for availability
Select an Option
34 mm
Select an Option
1 ml
Select an Option
25 ml
Select an Option
Select an Option
49 mm
Select an Option
01
Select an Option
1
About this item
PMP (TPX), crystal clear.
  • Good chemical resistance
  • Heat resistant up to +150 °C (up to +170 °C for short periods), autoclavable
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
Certifications: ISO 7056.
Frequently Bought Together