ContactUsLinkComponent
Beakers, low form
Beakers, low form
Catalog # 213-0196
Supplier:  KARTELL
Supplier #: 1806
undefined
Beakers, low form
Catalog # 213-0196
Supplier:  KARTELL
Supplier #: 1806
Log in for availability
Select an Option
87 mm
Select an Option
10 ml
Select an Option
500 ml
Select an Option
Select an Option
119 mm
Select an Option
05
Select an Option
1
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read moulded graduations.
ISO 7056
Certifications: ISO 7056
Frequently Bought Together