Optical bottom plates, 96-well, CC²™ coated, Nunc™
Catalog # 734-1354
Supplier: Thermo Fisher Scientific
Specifications
- Poč. jamek:96
- Doporučený pracovní objem:50 - 200 µl/well
- Sterilní:+
- Barva:Černá
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Bal.:30
- Objem jamky:400 µl
Specifications
About this item
PS with coverglass base, sterile, with lid. CC2™ surface treatment closely mimics a biological surface similar to Poly-D-Lysine and is a superior surface for attachment and growth of fastidious cells.
- White or black upper structure with coverglass bottom for minimum light scatter and low auto-fluorescence, ensuring accurate results due to higher signal-to-noise ratios
- Optimum clarity for viewing well contents