ContactUsLinkComponent
VWR® Capillary Tubes for Melting point Determinations
VWR® Capillary Tubes for Melting point Determinations
Catalog # 321242C
Supplier:  VWR Chemicals
undefined
VWR® Capillary Tubes for Melting point Determinations
Catalog # 321242C
Supplier:  VWR Chemicals
Specifications
  • Ø vněj.:
    ~1 mm
  • Popis:
    Capillary tubes, hard glass
  • Délka:
    100 mm
  • Bal.:
    100
Specifications
About this item
Hard glass.
  • Open both ends