Order Entry
Czech Republic
ContactUsLinkComponent
Minicon® Static Concentrators
Minicon® Static Concentrators
Katalogové číslo MILY9051
Dodavatel:  MILLIPORE
New Product
Číslo CAS:  
undefined
Minicon® Static Concentrators
Katalogové číslo MILY9051
Dodavatel:  MILLIPORE
New Product
Číslo CAS:  

Specifikace

  • Bal.:
    50 Tests
  • Popis:
    Minicon CS15, 10 cells/unit, Minimum final concentrate volume: 30 µl
  • Objem:
    2,5 ml

Specifikace

O tomto produktu

The Minicon® Static concentrators to concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis. The B15 unit has 8 cells and can hold up to 5 ml of sample. The CS15 has 10 cells and is suited for volumes up to 2,5 ml. The absorbent pulls solvent and salts through the ultrafilter, concentrating the sample.

  • Static concentrator, requiring no accessories
  • Absorbent pulls solvent and salts through ultrafilter, concentrating sample
  • Concentration factor up to 100X
  • Low hold-up volume
  • Nominal Molecular Weight Limit (NMWL): 15 kDa

Concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis (e.g., Bence Jones proteins in urine).

Applications: Concentration of urine and cerebrospinal fluid for enrichment of protein analytes (e.g., Bence Jones Proteins in Urine)