Orders LinkContactUsLinkComponent
Multiscreen® High Volume Filter Plates
Multiscreen® High Volume Filter Plates
Dodavatel:   MILLIPORE
Multiscreen® High Volume Filter Plates
Dodavatel:  MILLIPORE
Certifikáty
Multiscreen® High Volume Filter Plates
Multiscreen® High Volume Filter Plates
2 Dostupných variant níže
Product Details & Documents
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
  • Sample prep
  • Protein kinase/phosphatase assays
  • Receptor/ligand binding assays
Product Family Options
Informace o produktu
  • Sekvence
    Materiál
    Barva
    Membrána
    Velikost pórů
    Poč. jamek
    Sterilní
    Dostupnost
    Price
  • MILYMVHVN4525
    MultiScreen® High volume filter plates
    MultiScreen® High volume filter plates
    1
    PP
    Matná
    Hydrophilic Durapore®
    membrane
    0,45 µm
    96
    +
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 25 KS
    1 * 25 KS - 36 369,90 Kč
    36 369,90 Kč
    1 * 25 KS
       36 369,90 Kč
  • MILYMVFCN1225
    MultiScreen® High volume filter plates
    MultiScreen® High volume filter plates
    2
    PP
    Matná
    Glass fiber membrane
    1,2 µm
    96
    -
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 25 KS
    1 * 25 KS - 36 369,90 Kč
    36 369,90 Kč
    1 * 25 KS
       36 369,90 Kč