Aspiration station Y-shaped tubing connector
Katalogové číslo GILSF1077418
Dodavatel: GILSON
Specifikace
- Popis:Y-tubing connector
- Sekvence:03
- Bal.:1
Specifikace
O tomto produktu
The Gilson safe aspiration station Y-shaped tubing connector is for use with the Gilson safe aspiration station.
- Connect two handles to Gilson Safe Sapiration Station station
The connector enables you to connect two handles to the station, allowing two people to use the station simultaneously.