ContactUsLinkComponent
SURFLO® Winged Infusion Sets
SURFLO® Winged Infusion Sets
Katalogové číslo TERU8SV27ELF
Dodavatel:  TERUMO
New Product
SURFLO® Winged Infusion Sets
Katalogové číslo TERU8SV27ELF
Dodavatel:  TERUMO
New Product
Specifikace
  • Rozchod:
    27
  • Popis:
    Winged infusion set (regular wall needle), 13 mm (needle length), 203 mm (tubing length)
  • Bal.:
    50
Specifikace
O tomto produktu
SURFLO winged infusion sets feature a precisely honed needle with a thin wall design, ensuring easy penetration and continued access with a single insertion.
The soft, flexible interlocking wings readily conform to the body of the patient, while the angle of the wings helps position the needle and reduce the risk of vessel damage during venipuncture.