Developer Tray, PVC
Developer tray, Bílá, 620×765×110 mm, 530×675×100 mm
Katalogové číslo SEMA658
Dodavatel: SEMADENI
Přejít na:
Product Details & Documents
Made from white PVC, with grooved bottom, very sturdy and low-priced tray, acid-proof, temperature-resistant up to +60 °C.
Specifikace
- Š×H×V vnějš.:620×765×110 mm
- Barva:Bílá
- Popis:Developer tray
- Š×H×V vnitř.:530×675×100 mm