ContactUsLinkComponent
Perforated Shelf Cart
Perforated Shelf Cart
Catalog # PALBCY0020
Supplier:  Palbam
New Product
Perforated Shelf Cart
Catalog # PALBCY0020
Supplier:  Palbam
New Product
Specifications
  • Popis:
    Perforated shelf cart, 1357L×732W×1650H mm
  • Bal.:
    1
Specifications
About this item
This four perforated shelf cart has three side protective railing. Stainless steel 304L electro-polished construction. Smooth running cleanroom casters, two with brakes.