Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy vodivostní
Standard vodivosti 1.413 µS/cm (25°C; 0,01 mol/l; KCl)
Standard vodivosti 1.413 µS/cm (25°C; 0,01 mol/l; KCl)
Standard vodivosti 1.413 µS/cm (25°C; 0,01 mol/l; KCl)
Catalog # WTWA300575
Supplier:
WTW
CAS Number:
Certificates
Expand image
Standard vodivosti 1.413 µS/cm (25°C; 0,01 mol/l; KCl)
Catalog # WTWA300575
Supplier:
WTW
CAS Number:
Certificates
Specifications
Typ balení:
Glass bottle
Bal.:
1 L
Referenční teplota:
25 °C
Popis:
Conductivity control standard 1413 µS/cm (0,01 mol/l; KCl)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation