Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy vodivostní
Standard vodivosti 100.000 µS/cm (25°C; 0,85 mol/l; KCl)
Standard vodivosti 100.000 µS/cm (25°C; 0,85 mol/l; KCl)
Standard vodivosti 100.000 µS/cm (25°C; 0,85 mol/l; KCl)
Catalog # YSIA065274
Supplier:
YSI ENVIRONMENTAL
CAS Number:
Expand image
Standard vodivosti 100.000 µS/cm (25°C; 0,85 mol/l; KCl)
Catalog # YSIA065274
Supplier:
YSI ENVIRONMENTAL
CAS Number:
Specifications
Bal.:
950 mL
Referenční teplota:
25 °C
Popis:
Standard vodivosti 100.000 µS/cm (25°C; 0,85 mol/l; KCl)
Specifications
About this item
Product Details
Product Details