Sudan II, Supelco®
Glass bottle for solids, 25 mg, 2315
07937-25MG
: Merck
: 1-(2,4-Dimethylphenylazo)-2-naphthol, Solvent Orange 7, 1-((2,4-Dimethylphenyl)diazenyl)naphthalen-2-ol
- :3118-97-6
- :C₁₈H₁₆N₂O
- :MFCD00003896
- :276.34
- :156…158 °C
- :M-Clarity™ quality level = MQ100
- :Ambient
- :221-490-4
- Typ balení:Glass bottle for solids
- Bal.:25 mg
- Sekvence:2315