Temperature Data Logger for Process Validation, EBI 12-T240
O tomto produktu
The EBI 12–T240 is a precise temperature data logger for process validation in laboratories and the pharmaceutical industry. With a temperature range of –200 to +200 °C and an accuracy of up to 0,05 °C, it enables precise temperature measurements in critical applications such as sterilisation, disinfection and ovens.
- Suitable for steam sterilisers, washer–disinfectors
- Temperature range –200 to +200 °C
- High accuracy up to 0,05 °C
- One bendable metal probe (Ø 1,50 mm)
The EBI 12–T240 temperature data logger from ebro fulfils the stringent requirements of the pharmaceutical industry and laboratory applications. It is ideal for process validation in steam sterilisers, washer-disinfectors and other critical processes where precise temperature control is required. With a wide temperature range from –200 to +200 °C and an accuracy of up to 0,05 °C, it ensures reliable results. Thanks to real-time data transmission and flexible measurement interval selection, it is perfect for seamless documentation in validated processes. Its robust design ensures a long service life and high resistance in demanding environments.
Calibration points: –20 °C, 0 °C, +60 °C and +134 °C
Operating temperature: –90 to +150 °C
Dimensions: Ø 48×24 mm H, probe length 250 or 500 mm
Certifications: Certified according to ISO 17665, ISO 15883, (EC) 1935/2004.
Informace o dodávce: Includes 1×EBI 12-T240, 1×factory calibration certificate, manual.
2 Dostupných variant níže
Product Details & Documents
The EBI 12–T240 is a precise temperature data logger for process validation in laboratories and the pharmaceutical industry. With a temperature range of –200 to +200 °C and an accuracy of up to 0,05 °C, it enables precise temperature measurements in critical applications such as sterilisation, disinfection and ovens.
- Suitable for steam sterilisers, washer–disinfectors
- Temperature range –200 to +200 °C
- High accuracy up to 0,05 °C
- One bendable metal probe (Ø 1,50 mm)
The EBI 12–T240 temperature data logger from ebro fulfils the stringent requirements of the pharmaceutical industry and laboratory applications. It is ideal for process validation in steam sterilisers, washer-disinfectors and other critical processes where precise temperature control is required. With a wide temperature range from –200 to +200 °C and an accuracy of up to 0,05 °C, it ensures reliable results. Thanks to real-time data transmission and flexible measurement interval selection, it is perfect for seamless documentation in validated processes. Its robust design ensures a long service life and high resistance in demanding environments.
Calibration points: –20 °C, 0 °C, +60 °C and +134 °C
Operating temperature: –90 to +150 °C
Dimensions: Ø 48×24 mm H, probe length 250 or 500 mm
Certifications: Certified according to ISO 17665, ISO 15883, (EC) 1935/2004.
Informace o dodávce: Includes 1×EBI 12-T240, 1×factory calibration certificate, manual.
Documents
Data Loggers - On The Safe Side with Validated Processes
Poznámky k aplikaci
Professional Dataloggers
Produktová literatura
Specifications for Product Family
Specifikace
- Catalog No:
- EBRO1340-6611
- EBRO1340-6612
- Model:
- EBI 12-T240
- EBI 12-T241
- Snímač:
- PT1000, class A
Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 250 mm - PT1000, class A
Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 500 mm
- Ochranná třída:
- IP 68
- IP 68
- Intervaly záznamu:
- 250 ms to 24 hrs.
- 250 ms to 24 hrs.
- Hmotnost:
- 90 g
- 90 g
- Datová paměť - Počet odečtů:
- 100,320
- 100,320
- Baterie:
- Lithium, 2× BR1225A, 3 V user–replaceable
- Lithium, 2× BR1225A, 3 V user–replaceable
Specifikace
Product Family Options
Data loggers
Informace o produktu
- SekvenceRozsahPřesnostRozlišeníDostupnostPrice
- EBRO1340-6611EBI 12-T240, high temperature data logger with two bendable probes, 250 mm lengthEBI 12-T240, high temperature data logger with two bendable probes, 250 mm length01–200…+200 °C±1,5 °C (–200…–90 °C)
±0,5 °C (–90…–40 °C)
±0,2 °C (–40…0 °C)
±0,1 °C (0…+120 °C)
±0,05 °C (+120…+140 °C)
±0,1 °C (+140…+150 °C)
±0,5 °C (+150…+200 °C)0,01 °CPřihlásit se pro dostupnost1 * 1 KS92 550,00 KčSpecifikace:More
Specifications
Cat. No.EBRO1340-6611ModelEBI 12-T240SnímačPT1000, class A
Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 250 mmOchranná třídaIP 68Intervaly záznamu250 ms to 24 hrs.Hmotnost90 gDatová paměť - Počet odečtů100,320BaterieLithium, 2× BR1225A, 3 V user–replaceable - EBRO1340-6612EBI 12-T241, high temperature data logger with two bendable probe, 500 mm lengthEBI 12-T241, high temperature data logger with two bendable probe, 500 mm length02–200…+200 °C±1,5 °C (–200…–90 °C)
±0,5 °C (–90…–40 °C)
±0,2 °C (–40…0 °C)
±0,1 °C (0…+120 °C)
±0,05 °C (+120…+140 °C)
±0,1 °C (+140…+150 °C)
±0,5 °C (+150…+200 °C)0,01 °CPřihlásit se pro dostupnost1 * 1 KS92 550,00 KčSpecifikace:More
Specifications
Cat. No.EBRO1340-6612ModelEBI 12-T241SnímačPT1000, class A
Radial, 1 external temperature probe, bendeble, Ø 1,5 mm, 500 mmOchranná třídaIP 68Intervaly záznamu250 ms to 24 hrs.Hmotnost90 gDatová paměť - Počet odečtů100,320BaterieLithium, 2× BR1225A, 3 V user–replaceable
Software and readout station
Informace o produktu
- Katalogové čísloSekvencePopisDostupnostPrice
- 13SI 1200, 4-port Interface IF 200, Software Winlog.proPřihlásit se pro dostupnost1 * 1 KS77 220,00 Kč
Complementary Products
Informace o produktu
- Katalogové čísloPopisSekvenceRozlišeníPřesnostRozsahDostupnostPrice
- EBRO1340-6610EBI 12-T222 Temperature logger, 1-channel, radial, 40 mm rigid metal probe34Přihlásit se pro dostupnost1 * 1 KS - 49 020,00 Kč
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12Battery replacement set EBI 12; 3× battery, 6× O-ring, 1× unscrewing aid EBI 12EBRO1100-013029Přihlásit se pro dostupnost1 * 1 KS - 7 530,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Battery replacement set EBI 12; 3× battery, 6× O-ringBattery replacement set EBI 12; 3× battery, 6× O-ring620-16937Přihlásit se pro dostupnost1 * 1 KS - 5 280,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 1102, Inlay for data logger sets made of plastic for EBI 16 and EBI 12 data logger and interfaceAL 1102, Inlay for data logger sets made of plastic for EBI 16 and EBI 12 data logger and interfaceEBRO1248-110227Přihlásit se pro dostupnost1 * 1 KS - 5 280,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 128, Carrying case systainer 1,25 HUAL 128, Carrying case systainer 1,25 HUEBRO1248-012832Přihlásit se pro dostupnost1 * 1 KS - 5 400,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 1100, Inlay for data logger sets made of plastic for EBI 12 data logger and interfaceAL 1100, Inlay for data logger sets made of plastic for EBI 12 data logger and interfaceEBRO1248-110025Přihlásit se pro dostupnost1 * 1 KS - 5 280,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- EBI NI - 140, Grommets for fixing sensorsEBI NI - 140, Grommets for fixing sensorsEBRO1340-198815Přihlásit se pro dostupnost1 * 100 KS - 7 530,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 1103, Inlay for data logger sets made of plastic for EBI 12 data logger and interfaceAL 1103, Inlay for data logger sets made of plastic for EBI 12 data logger and interfaceEBRO1248-110328Přihlásit se pro dostupnost1 * 1 KS - 5 280,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 102, Silicone protection for EBI 12 radial, 2 channelsAL 102, Silicone protection for EBI 12 radial, 2 channelsEBRO1340-602224Přihlásit se pro dostupnost1 * 1 KS - 7 110,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 190, Silicone cable strap set consisting of 10 pcsAL 190, Silicone cable strap set consisting of 10 pcsEBRO1248-019033Přihlásit se pro dostupnost1 * 1 KS - 1 500,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 1101, Inlay for data logger sets made of plastic for EBI 12 data loggerAL 1101, Inlay for data logger sets made of plastic for EBI 12 data loggerEBRO1248-110126Přihlásit se pro dostupnost1 * 1 KS - 5 280,00 Kč
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 121, Opening tool for EBI 12
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- EBI-KV3, Compression fitting
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- AL 122, Eyelet for EBI 12 Data logger consisting of 1 eyelid and 2 sealing O-ringsAL 122, Eyelet for EBI 12 Data logger consisting of 1 eyelid and 2 sealing O-ringsEBRO1248-012231Přihlásit se pro dostupnost1 * 1 KS - 1 470,00 Kč