Orders LinkContactUsLinkComponent
Orbipac CPS51 pH-ORP Sensor
pH electrode including and excluding integrated NTC temperature sensor, Fibre, Handheld and laboratory measuring devices
Katalogové číslo ENDRCPS51-1EK2FDA
Dodavatel:   Endress+Hauser
Orbipac CPS51 pH-ORP Sensor
Katalogové číslo ENDRCPS51-1EK2FDA
Dodavatel:  Endress+Hauser
Orbipac CPS51 pH-ORP Sensor
pH electrode including and excluding integrated NTC temperature sensor, Fibre, Handheld and laboratory measuring devices
Přejít na:
Price
8 610,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
pH electrode for handheld and laboratory measuring devices.
  • Fixed cable: 1,5 m
  • CPS51 excluding temperature sensor: 1EK2FDA
  • Working limit: Pressureless
Certifications: EN IEC 63000:2018.
Specifikace
  • Určeno pro:
    Handheld and laboratory measuring devices
  • Popis:
    pH electrode including and excluding integrated NTC temperature sensor
  • Diafragma:
    Fibre