Orders LinkContactUsLinkComponent
hMSC Proliferation Medium, Basal
hMSC Proliferation Medium, Basal
Dodavatel:   PROVITRO
hMSC Proliferation Medium, Basal
Dodavatel:  PROVITRO
Certifikáty
hMSC Proliferation Medium, Basal
2 Dostupných variant níže
Product Details & Documents
The hMSC Proliferation Medium is a sterile liquid culture medium designed for cultivating human mesenchymal stem cells (hMSC). Delivered as a basal medium, it is well-suited for culturing hMSC upon the addition of supplement mix components.
  • Formulation is optimised for the initial seeding of 6000 cells/cm², supporting growth up to confluence (approximately 90%)
  • Medium does not require feeder layers, matrix substrates, or additional substances for effective cell cultivation
Due to the possibility of reduced proliferative activity, we recommend to use the antibiotic supplement for freshly isolated cells only. Endothelial cell proliferation medium is thoroughly tested after each production. All components are tested in a stringent biological assay.
Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency, and the population doubling time.
Storge: Store the supplemented Skeletal muscle cell growth medium in a dark environment at temperatures ranging from 4 to 8 °C for a maximum duration of one month. Avoid heating the medium beyond 37 °C and refrain from using uncontrolled heat sources, such as microwave appliances. When utilising a portion of the medium, extract the required amount from the bottle and warm it appropriately. Never freeze the medium, as this could result in elevated salt concentrations due to the separation of pure water during freezing, leading to irreversible damage.
Specifications for Product Family
Specifikace
Specifikace
  • Catalog No:
  • PRTV2000901
  • PRTV2110901
  • Media Base:
  • Basal Medium
  • Basal Medium
Specifikace
Product Family Options
Informace o produktu
  • Sekvence
    Formát média
    Bal.
    Dostupnost
    Price
  • PRTV2000901
    hMSC proliferation medium, basal
    hMSC proliferation medium, basal
    01
    Kapalný
    500 mL
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 500 mL
    1 * 500 mL - 4 200,00 Kč
    4 200,00 Kč
    1 * 500 mL
       4 200,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    PRTV2000901
    Media Base
    Basal Medium
  • PRTV2110901
    hMSC proliferation medium, FCS-kit
    hMSC proliferation medium, FCS-kit
    02
    Kapalný
    500 mL
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 500 mL
    1 * 500 mL - 7 150,00 Kč
    7 150,00 Kč
    1 * 500 mL
       7 150,00 Kč
    Specifikace:
    More
    Specifications
    Cat. No.
    PRTV2110901
    Media Base
    Basal Medium