Pierce™ Blocking Buffers, Protein Free
PIER37570
:
- Bal.:1 L
- Popis:Pierce™ protein- free (TBS) blocking buffer
- Sekvence:04
These protein-free blocking buffers are either a PBS or a TBS formulation of a non protein compound that provides effective blocking for membrane based and plate based protein detection methods, resulting in extremely low background.
- Streptavidin friendly that is absolutely free of biotin
- Effective for all kinds of protein detection systems
- Optimized and validated in many protein methods to provide high signal-to-noise ratio
- Protein free blocker that minimizes or eliminates cross reactivity associated with protein-based blocking buffers
These buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, western blotting, glass slide arrays and other applications.
: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store product at room temperature. After opening, store product at 4˚C.