AmberChrom™ chloride form 1x2 50-100 mesh
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:69011-19-4
:MFCD00132718
Formula:(C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
MDL Number:MFCD00146763
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to yellow beads
- Total exchange capacity:≥0.7 eq/l (Cl form)
- Water retention capacity:65 to 75 % (Cl form)
- Particle size:50 to 100 mesh
Specification Test Results
Frequently Bought Together