Anti-Rat IgG (H+L) Goat Polyclonal Antibody (HRP)
Catalog # LEIC2AH-015
Supplier: LEICA MICROSYSTEMS
New Product
Specifications
- Název antigenu:Rat IgG (H+L)
- Klonovalita:Polyclonal
- Spojení:HRP (Horseradish Peroxidase)
- Hostitel:Goat
- Izotyp:IgG
- Čistota:Affinity purified antibody is >95% based on SDS-PAGE
- Skladovací pufr:TBS, 1% BSA, 0,1% proclin 150, 10% heat-inactivated animal serum
- Skladovací teplota:2...8 °C
- Imunogen:Purified Rat IgG, whole molecule
- Bal.:15 mL
Specifications
About this item
Type:
Antigen: Rat IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype: IgG
Reactivity: