BasicKjel Distillation Unit
Katalogové číslo BUCH11K36521110
Dodavatel: BUCHI
New Product
Specifikace
- Popis:BasicKjel distillation unit with glass protection vessel and without chiller
- Environmentally Preferable:
- Bal.:1
Specifikace
O tomto produktu
For protein and nitrogen determination (TKN, NPN, TVBN, ammonia, nitrate/nitrite and casein). Possibility to connect Metrohm or SI analytics titrators for the MultiKjel.
Three possible equipment:
EasyKjel: Basic unit
BasicKjel: Intermediate unit
MultiKjel: Advanced unit
Two types of protection: Plastic or glass protective vase: To protect the condenser from splashes caused by the distillation of water vapour.