O tomto produktu
AlphaTec® 04-005 Gloves are heavyweight PVC supported glove with better comfort, robust, long-cuffed PVC gloves, offering grip, comfort and chemical resistance.
- Protection from varied chemical hazards
- Suitable food safety gloves for multiple applications
- A balance of hand protection and comfort
Heightened protection: With their special PVC and nitrile formulation, AlphaTec 04-005 protective gloves blend high-level chemical resistance abrasion protection and sustained flexibility, even at low temperatures.
Food handling compliance: These latex-free industrial gloves also satisfy FDA and EU food processing and hygiene norms, resulting in ideal food safety gloves for a variety of applications.
Increased comfort: Thanks to their 13-gauge seamless cotton design, they offer an excellent balance between personal protection and user comfort.
Enhanced features: These AlphaTec gloves foam grip design guards against oil and grease, while maintaining superb grip in wet and oily environments, and their 380 mm glove length further enhances wearer protection.
Advanced ergonomics: In addition, their ergonomic hand shape further enhances wearer comfort and makes them easy to use over a prolonged period.
Applications are all food processing applications that have food contact, chemical handling, maintenance, cleaning.
Specifikace
- Catalog No:
- ANSE04005/9
- ANSE04005/8
- ANSE04005/10
- ANSE04005/11
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Materiál:
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- Provedení:
- Foam
- Foam
- Foam
- Foam
- Typ manžety:
- Rukavice
- Rukavice
- Rukavice
- Rukavice
- Tloušťka drapáku:
- 1,80 mm
- 1,80 mm
- 1,80 mm
- 1,80 mm
- Antistatický:
- +
- +
- +
- +
- Schváleno pro styk s potravinami:
- +
- +
- +
- +