Magnetic Stirrer with Stand
Katalogové číslo METR28010040
Dodavatel: METROHM
New Product
Specifikace
- Popis:Magnetic stirrer with stand
- Bal.:1
Specifikace
O tomto produktu
Magnetic stirrer including base plate, support rod and electrode holder for supplementing the Titrino plus, Dosimat plus, titrandos, sample processors, 805 Dosimat and 780/781 pH meters as well as the 856 and 867 measuring modules.
With permanently attached cable for MSB (Metrohm Serial Bus).