Order Entry
Czech Republic
ContactUsLinkComponent
NIST® Green Tea Containing Solid Oral Dosage Form
NIST® Green Tea Containing Solid Oral Dosage Form
Katalogové číslo SIALNIST3256
Dodavatel:  Merck
New Product
Číslo CAS:  
NIST® Green Tea Containing Solid Oral Dosage Form
Katalogové číslo SIALNIST3256
Dodavatel:  Merck
New Product
Číslo CAS:  

Specifikace

  • Bal.:
    1 SET
  • Popis:
    Green tea containing solid oral dosage with the format of matrix material
  • Pk Info:
    5×2,5 g

Specifikace

O tomto produktu

Application: The SRM is primarily intended to validate analytical methods used to determine catechins, xanthines, andtoxic elements present in solid oral dosage forms in cluding green tea and similar matrices. Additionally, it is also suitable for ensuring quality while assigning values to in-house control materials.

  • The certified value of the SRM was determined by calculating the mass fraction values, which were obtained through two methods
  • Ultraviolet absorbance detection (UV)
  • Mass spectrometry (MS)
  • Certified and reference mass fraction values for catechin, gallocatechin, gallic acid, theanine, and other elements are provided on a dry-mass basis and values are metrologically traceable to the SI unit of mass expressed as micrograms per gram
  • The NIST certificate is provided with expiration certificate, storage, handling, use and maintenance instructions

Grade: Certified reference material
Quality level: 100

Packaging: One unit of SRM 3256 contains five packets of powdered material, each containing approximately 2,5 g.