ContactUsLinkComponent
Vacuum Filtration Assembly
Vacuum Filtration Assembly
Catalog # SIALZ290440
Supplier:  Merck
New Product
Vacuum Filtration Assembly
Catalog # SIALZ290440
Supplier:  Merck
New Product
Specifications
  • Membrána:
    Glass
  • Popis:
    Vacuum filtration assembly, for 47 mm filters with glass support, #8 stopper, without filtration flask, neck Ø: 41 mm
  • Bal.:
    1
Specifications
About this item
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications.
  • With graduation
  • Glass housing
Clamp seals filter membrane between flanges on funnel top and funnel stem.
Delivery: Includes: Funnel top, funnel stem/membrane support, aluminium clamp, stopper (except standard taper units) and filtration flask.