Vacuum Filtration Assembly
Catalog # SIALZ290440
Supplier: Merck
New Product
Specifications
- Membrána:Glass
- Popis:Vacuum filtration assembly, for 47 mm filters with glass support, #8 stopper, without filtration flask, neck Ø: 41 mm
- Bal.:1
Specifications
About this item
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications.
- With graduation
- Glass housing
Clamp seals filter membrane between flanges on funnel top and funnel stem.
Delivery: Includes: Funnel top, funnel stem/membrane support, aluminium clamp, stopper (except standard taper units) and filtration flask.