Ultrasonic generator, GM 5200
Katalogové číslo BAND3736
Dodavatel: BANDELIN-ELECTRONIC
New Product
Specifikace
- Popis:Ultrasonic generator GM 5200, external dimensions: 380×195×215 mm, 230 V EU plug
- Délka:1.5 m
- Bal.:1
- Určeno pro:UW 5100, UW 5200
Specifikace
O tomto produktu
There are two different ultrasonic generators to choose from. They are specified for various applications due to their different power ranges. The easy-care and robust plastic housing, equipped with a practical recessed grip for easy transportation and positioning on the laboratory bench, is identical for both versions.
The various SONOPULS ultrasonic homogenisers in the HD 5000 series can be connected directly to the ultrasonic generators depending on their compatibility. Mains connection 230 VAC (±10 %), 50/60 Hz.
The modern 7" touch display offers intuitive, user-friendly operation. Setting the set values for amplitude, pulsation and time and the display of the actual values enable reproducible results.