Titrator, Volumetric, 901 Titrando
Catalog # METR29010010
Supplier: METROHM
New Product
Specifications
- Popis:Titrator
- Pro použití s:OMNIS software, tiamo software, or touch control unit
- Bal.:1
Specifications
About this item
The Karl Fischer Titrando for the modern titration laboratory. In addition to the Karl Fischer mode (KFT), the high-end titrator is also equipped with the SET mode and can thus perform titrations to a predefined endpoint.
'iTrode ready': The high-resolution, galvanic measuring interface - now also for the first time with additional digital measuring input for the intelligent. 'iTrodes' electrodes: It is additionally possible to perform pH measurements with a maximum of precision.
For use with OMNIS Software, tiamo software, or Touch Control unit. Compliance with GMP/GLP and FDA regulations such as 21 CFR Part 11, if required.