Titrator, Coulometeric, 851 Titrando
METR28510110
: METROHM
New Product
- Popis:851 Titrando with generator electrode without diaphragm
- Bal.:1
Coulometer including generator electrode without diaphragm.
Coulometry is the ideal method for water content determination in liquids, solids, and gases when it comes to water content determination in the trace range (0,01 to 10 mg absolute water). In addition, coulometry is an absolute method and thus no titer determination is necessary.
Coulometric titrations are carried out easily and quickly with the 851 Titrando.
Recommended measuring range: 0,01 - 200 mg absolute water.