Digi-Sense High-Temperature Thermocouple Probe
Katalogové číslo COCO08516-96
Dodavatel: CONTROL
New Product
Specifikace
- Rozsah:−250...1,100 °C
- Popis:Digi-Sense high-temperature thermocouple probe, grounded
- Délka:30,48 cm
- Materiál:Inconel® slitina nikl-chrom-železo 601
- Připojení:Mini connector
- Bal.:1
- Typ:Type-K
Specifikace
O tomto produktu
This Digi-Sense Type-K High-Temperature Thermocouple Probe can withstand the harsh demands of industrial environments.
- Delivers fast, accurate temperature readings
- Probe is flexible and can be bent around pipe diameters that are twice the probe sheath diameter
- Response time is 0,85 seconds or greater
- Braided cable with fiberglass insulation to withstand high temperatures
- Sheath made from ultra-durable Inconel® 600
- Includes a color-coded ANSI Type-K mini connector
- Applications: Industrial environments
The cable features fiberglass insulation, allowing it to perform well in high temperatures. Because the cable is flexible, it's easy to wrap it around pipes and get the fast, accurate measurements your facility depends on. Note: Temperature range is for the probe tip only.