CrossLab CS Gas Leak Detector
Handheld electronic gas leak detector
Katalogové číslo HEWLG6693A
Dodavatel: Agilent
Přejít na:
Product Details & Documents
Leak detector will detect leaks or verify leak-free tubing and fittings for various gas types, including hydrogen, nitrogen, helium, and others, throughout the lab or site.
- Unique design
- Used for other analytical techniques like LC/MS and ICP-MS
The cartridge design combines the two most critical GC flow path monitoring tasks, including flow rate and leak detection monitoring, into a single handheld system with the bundle offering.
Specifikace
- Popis:Handheld electronic gas leak detector