M-Enterococcus Agar Base Dehydrated Culture Media, HiMedia
HIMEM1108-500G
: HIMEDIA
New Product
:
- Bal.:1
- Popis:M-Enterococcus agar base, 500 g
For the isolation and enumeration of enterococci in water, sewage, food and other materials by membrane filter technique as well as direct plating of specimens.
- Microbiological culture media
- 500 g poly bottles