Order Entry
Czech Republic
ContactUsLinkComponent
 
NIST® Organic Contaminants in House Dust
  SIALNIST2585
 :  Merck
New Product
 :  
NIST® Organic Contaminants in House Dust
  SIALNIST2585
 :  Merck
New Product
 :  

 

  • Bal.:
    10 g
  • Popis:
    Organic contaminants in house dust
  • Tvar:
    Matrix material

 

 

This Standard reference material (SRM) is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polybrominated diphenyl ether (PBDE) congeners in house dust and similar matrices. All of the constituents for which certified, reference, and information values are provided in SRM 2585 were naturally present in the dust material before processing.

  • Grade: Certified reference material
  • Quality level: 100

Application: Industrial hygiene, laboratory chemicals, manufacture of substances, innovative non-targeted screening approach using high-resolution mass spectrometry, endocrine disrupting potential of total and bioaccessible extracts of dust, comprehensive identification and ubiquitous occurrence of rubber-derived vulcanisation accelerators in urban dusts.

 : A unit of SRM 2585 consists of one bottle containing approximately 10 g of house dust.