TASKI® Jontec No1 F1c Heavy-Duty Floor Stripper - No Rinse
Katalogové číslo DIVY101102722
Dodavatel: Diversey
New Product
Specifikace
- Kapacita:2×5 L
- Popis:F1c Heavy-duty floor stripper - no rinse
- Bal.:2 Can
Specifikace
O tomto produktu
Heavy duty stripper for effective removal of floor emulsions and maintenance films from alkali resistant hard floors.
- Highly alkaline
- High wetting and emulsifying power
- Fast and easy emulsion removal without rinsing
Unlike conventional strippers, its revolutionary formulation enables floors to be ready for re-coating without the need to rinse, reducing normal stripping time by up to 50%.