SO Green™ 520WS Singlet Oxygen Sensor *Water-Soluble for Extracellular Applications*
Catalog # AATB16061
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Bal.:100 µG
- Popis:SO Green™ 520WS Singlet Oxygen sensor *Water-soluble for extracellular applications*, 10×100 μg
Specifications
About this item
SO Green™ 520WS Singlet Oxygen Sensor has been developed as a water-soluble fluorescent probe for detecting extracellular singlet oxygen since it is cell-impermeant.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.