ROX Dye qPCR Calibration Solution *10000X*
Catalog # AATB67034
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Bal.:100 µl
- Popis:ROX Dye qPCR calibration solution *10000X*
Specifications
About this item
ROX dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.