HIS Lite™ iFluor® 647 Tris NTA Chelator
Catalog # AATB12658
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Bal.:100 µG
- Popis:HIS Lite™ iFluor™ 647 Tris NTA chelator
Specifications
About this item
iFluor® 647 Tris-NTA Chelator is used as a sensitive red fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion such as Ni²⁺ in cells, solution and solid surfaces.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.