FluoroQuest™ Antifade Mounting Medium
AATB20007
New Product
:
- Bal.:5 mL
- Popis:FluoroQuest™ Antifade mounting medium
FluoroQuest™ Antifade mounting medium is a specialised and stable formula that effectively preserves fluorescence by preventing rapid photobleaching of fluorescent proteins and dyes.
- Stable for 12 months
Quality tested in-house.
: For research use only.