Buccutite™ Rapid PerCP Antibody Labelling Kit
AATB5407
New Product
:
- Bal.:2
- Includes:2 labellings
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PerCP antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PerCP, APC, PE and iFluor® tandem dyes.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
: Shipped with blue ice.
: Research use only.