Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Katalogové číslo AATB1302
Dodavatel: AAT BIOQUEST
New Product
Číslo CAS:
Specifikace
- Bal.:2
- Includes:2 labellings
Specifikace
O tomto produktu
Microscale optimised for labelling 100 µg antibody per reaction. Many biological compounds present in cells, serum, or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Robust protocol
- Consistent results
- 12-month shelf life
Designed to provide high quality labelled antibodies for multiple downstream applications.
Delivery: Shipped at RT.
Caution: Research use only.