ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
Catalog # AATB17300
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Bal.:500 Tests
- Popis:ReadiView™ blue real-time PCR visualisation dye *200X*
Specifications
About this item
ReadiView™ Blue Real-Time PCR Visualisation Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.